Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Photogate
SDPSCI_ED

Catalog No. s16279nd Shop All Vernier Software Products
Click to view available options
Grade:
K-8, High School, College

Includes

Vernier Photogate, (1) Interface cable, (1) Accessory rod

Requires

Requires an interface and software, sold separately

Use the Photogate to study free fall, rolling objects, collisions, and pendulums.

Power req: 5 VDC at 40 mA Infrared source: Peak at 880 nm Dimension: 75 mm gate width Output is high and LED off for unblocked gate Output is low and LED on for blocked gate Beam approximately 9 mm from the end of the gate arms

Warranty and Services

5-year limited warranty

Specifications

Grade K-8, High School, College
Interface Required
Warranty 5 Year Limited
For Use With (Application) Educational use only
Width (Metric) 75 mm (Gate)
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.