Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHOX2B Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB343844 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343844 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343844 Supplier Novus Biologicals Supplier No. NBP241179PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PHOX2B Polyclonal antibody specifically detects PHOX2B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen PHOX2B
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias NBPHOX, NBPhoxPhox2b, Neuroblastoma Phox, paired mesoderm homeobox 2b, paired mesoderm homeobox protein 2B, paired-like homeobox 2bNBLST2, PHOX2B homeodomain protein, PMX2Bneuroblastoma paired-type homeobox protein
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the center of human PHOX2B. The immunogen is located within amino acids 130 - 180 of PHOX2B. Amino Acid Squence: FQRNRRAKFRKQERAA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 8929
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.