Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHRF1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP118933 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP118933 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP118933 Supplier Novus Biologicals Supplier No. NBP118933
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PHRF1 Polyclonal specifically detects PHRF1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen PHRF1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q9P1Y6
Gene Alias CTD binding SR like protein rA9, CTD-binding SR-like protein rA9, KIAA1542RNF221, PHD and RING finger domain-containing protein 1, PHD and ring finger domains 1
Gene Symbols PHRF1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1400 and 1450 of human PHD and ring finger domains 1 using the numbering given in entry NP_065952.1 (GeneID 57661).
Molecular Weight of Antigen 179 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57661
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.