Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pht1 / Histone H2A.Zac (pan-acetyl) Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
$156.65 - $620.58
Specifications
Antigen | Pht1 / Histone H2A.Zac (pan-acetyl) |
---|---|
Concentration | Lot-Specific |
Applications | Dot Blot, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2756
![]() ![]() |
Active Motif
39643 |
10 μL |
Each for $156.65
|
|
|||||
50-199-2755
![]() ![]() |
Active Motif
39642 |
100 μL |
Each for $620.58
|
|
|||||
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene encodes a member of the histone H2A family and generates two transcripts through the use of the conserved stem-loop termination motif, and the polyA addition motif.Specifications
Pht1 / Histone H2A.Zac (pan-acetyl) | |
Dot Blot, Western Blot | |
Unconjugated | |
Rabbit | |
Yeast | |
Q12692 | |
HTZ1 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Lot-Specific | |
Polyclonal | |
Liquid | |
RUO | |
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8 | |
854150 | |
This Pht1 / H2AZ acetyl antibody was raised against a peptide containing acetyl-lysine at positions 5, 7, 12 and 16 from the amino terminus of Pht1, the histone H2AZ variant in S. pombe. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
HTZ1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title