Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phusion™ High-Fidelity DNA Polymerase PROMO

A gold standard for high performance PCR.

Supplier:  Thermo Scientific™ F530L

 View more versions of this product

Catalog No. F530LW25


Description

Description

Features of Phusion™ High-Fidelity DNA Polymerase (2 U/μL) include:

  • High fidelity (52X Taq)
  • Fast PCR due to short extension times (15–30 s/kb)
  • Robust performance, minimal optimization needed
  • High yields of PCR products with minimal enzyme amounts
  • Available in Green buffer format for direct loading of PCR products on gels (F-534S or F-534L)

Applications

  • High-fidelity PCR
  • Cloning
  • Template generation for sequencing
  • Amplification of difficult (GC-rich) templates
  • Long-range PCR (up to 20 kb)
  • Mutagenesis
  • High throughput PCR
  • Microarray

Note: Annealing rules for Phusion DNA polymerases are different from many common DNA polymerases (such as Taq DNA polymerases). For optimal results, use our convenient Tm calculator.

Specifications

Specifications

2 U/μL
High
Fast
500 Units
Standard PCR, High-fidelity PCR
No
Blunt
20 kb or less
Phusion DNA Polymerase (2 U/μL), 5X Phusion HF GC Buffers, DMSO, 50 mM MgCl2 Solution, Phusion HF Buffer and Phusion GC Buffers Each Provide 1.5 mM MgCl2 in the Final 1X Concentration. Store at -20°C.
Phusion High-Fidelity DNA Polymerase
High-Fidelity DNA Polymerase
Dry Ice
52X
500 Reactions
Standalone
Colorless
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.