Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Phusion™ High-Fidelity DNA Polymerase PROMO
A gold standard for high performance PCR.
Specifications
Concentration | 2 U/μL |
---|---|
Content And Storage | Phusion DNA Polymerase (2 U/μL), 5X Phusion HF GC Buffers, DMSO, 50 mM MgCl2 Solution, Phusion HF Buffer and Phusion GC Buffers Each Provide 1.5 mM MgCl2 in the Final 1X Concentration. Store at -20°C. |
GC-Rich PCR Performance | High |
Polymerase | Phusion High-Fidelity DNA Polymerase |
Reaction Speed | Fast |
Description
Features of Phusion™ High-Fidelity DNA Polymerase (2 U/μL) include:
- High fidelity (52X Taq)
- Fast PCR due to short extension times (15–30 s/kb)
- Robust performance, minimal optimization needed
- High yields of PCR products with minimal enzyme amounts
- Available in Green buffer format for direct loading of PCR products on gels (F-534S or F-534L)
Applications
- High-fidelity PCR
- Cloning
- Template generation for sequencing
- Amplification of difficult (GC-rich) templates
- Long-range PCR (up to 20 kb)
- Mutagenesis
- High throughput PCR
- Microarray
Note: Annealing rules for Phusion DNA polymerases are different from many common DNA polymerases (such as Taq DNA polymerases). For optimal results, use our convenient Tm calculator.
Specifications
2 U/μL | |
High | |
Fast | |
Dry Ice | |
52X | |
Blunt | |
20 kb or less |
Phusion DNA Polymerase (2 U/μL), 5X Phusion HF GC Buffers, DMSO, 50 mM MgCl2 Solution, Phusion HF Buffer and Phusion GC Buffers Each Provide 1.5 mM MgCl2 in the Final 1X Concentration. Store at -20°C. | |
Phusion High-Fidelity DNA Polymerase | |
High-Fidelity DNA Polymerase | |
Standard PCR, High-fidelity PCR | |
No | |
Standalone | |
Colorless |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title