Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phycoerythrin Antibody (PE-DLF) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP20014655 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP20014655 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP20014655 Supplier Novus Biologicals Supplier No. NBP200146500UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

phycoerythrin Monoclonal specifically detects phycoerythrin in Non-species specific samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen phycoerythrin
Applications Flow Cytometry
Classification Monoclonal
Clone PE-DLF
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:1000
Formulation Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
Gene Alias PE, R-PE
Host Species Mouse
Immunogen The immunogen for this antibody is Phycoerythrin.
Purification Method Affinity Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4C. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.