Learn More
Eisco™ Phyllite Specimen
SCI_ED
Catalog No. S24788 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24788 1 Ea. 2 to 3 cm
S26637 10/Pk. 2 to 3 cm
S24789 12/Pk. 2 to 3 cm
S26681 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S24788 Supplier Eisco™ Supplier No. ESNG0059
Add to Cart
Add to Cart
Phyllite Specimen for demonstrations of metamorphic properties, such as glossy sheen and dense, foliated minerals.
  • Example of low-grade metamorphism from easily recognizable parent rock
  • For use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Fissile property of slate is retained, but has attained glossy sheen and soapy feel more indicative of metamorphic material
Specifications
Product Type Metamorphic Rock
For Use With (Application) Demonstration of metamorphic properties, such as glossy sheen and dense, foliated minerals
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Specimen Type Phyllite