Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI 3-Kinase p110 beta/PIK3CB Antibody (5A9), Novus Biologicals™
SDP

Catalog No. NB298210 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298210 50 μL
NB298211 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298210 Supplier Novus Biologicals Supplier No. NBP32638350UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PI 3-Kinase p110 beta/PIK3CB Monoclonal antibody specifically detects PI 3-Kinase p110 beta/PIK3CB in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen PI 3-Kinase p110 beta/PIK3CB
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 5A9
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunoprecipitation 1:200-1:1000
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias P110BETA, PI3K, PI3KBETA, PIK3C1, PIK3CB phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit beta
Host Species Rabbit
Immunogen A synthesized peptide derived from Human PI 3-Kinase p110 beta/PIK3CB [UniProt P42338]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5291
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.