Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI 3-Kinase p110 beta/PIK3CB Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB085780 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB085780 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB085780 Supplier Novus Biologicals Supplier No. NBP299597AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PI 3-Kinase p110 beta/PIK3CB Polyclonal specifically detects PI 3-Kinase p110 beta/PIK3CB in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen PI 3-Kinase p110 beta/PIK3CB
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias P110BETA, PI3K, PI3KBETA, PIK3C1, PIK3CB phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit beta
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human PI 3-Kinase p110 beta/PIK3CB.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5291
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.