Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI 3-Kinase p110 delta Antibody (OTI1D11), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP273396FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273396FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273396FR Supplier Novus Biologicals Supplier No. NBP273396FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PI 3-Kinase p110 delta Monoclonal specifically detects PI 3-Kinase p110 delta in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen PI 3-Kinase p110 delta
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI1D11
Conjugate DyLight 680
Dilution Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 2.7.1, EC 2.7.1.153, p110D, P110DELTA, Phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit delta, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta isoform, phosphoinositide-3-kinase C, phosphoinositide-3-kinase, catalytic, delta polypeptide, PI3K, PI3K-delta, PI3-kinase p110 subunit delta, PI3-kinase subunit delta, PtdIns-3-kinase subunit delta, PtdIns-3-kinase subunit p110-delta
Gene Symbols PIK3CD
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 286-610 of human PI 3-Kinase p110 delta (NP_005017) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Autophagy, mTOR Pathway, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5293.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.