Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI 3-Kinase p110 gamma Antibody (OTI2B1), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N273397A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273397A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273397A700 Supplier Novus Biologicals Supplier No. NBP273397AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PI 3-Kinase p110 gamma Monoclonal specifically detects PI 3-Kinase p110 gamma in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen PI 3-Kinase p110 gamma
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2B1
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 1-phosphatidylinositol 3-kinase, EC 2.7.1, EC 2.7.1.153, p110-gamma, p120-PI3K, phosphatidylinositol 3 kinase gamma, p110 gamma, phosphatidylinositol 3-kinase catalytic 110-kD gamma, phosphatidylinositol 3-kinase, catalytic, gamma polypeptide, Phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit gamma, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma isoform, phosphoinositide-3-kinase gamma catalytic subunit, phosphoinositide-3-kinase, catalytic, gamma polypeptide, PI3CG, PI3K, PI3Kgamma, PI3K-gamma, PI3-kinase subunit gamma, PIK3, PtdIns-3-kinase subunit gamma, PtdIns-3-kinase subunit p110-gamma
Gene Symbols PIK3CG
Host Species Mouse
Immunogen Full length recombinant protein of human PI 3-Kinase p110 gamma (NP_002640) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline mTOR Pathway, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5294.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.