Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI 3-Kinase p55 gamma Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$480.74

Specifications

Antigen PIK3R3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP158332
SDP
View Documents
Novus Biologicals
NBP158332
100 μL
Each of 1 for $480.74
Only null left
Add to Cart
 
Description

Description

PI 3-Kinase p55 gamma Polyclonal specifically detects PI 3-Kinase p55 gamma in Human samples. It is validated for Western Blot.
Specifications

Specifications

PIK3R3
Polyclonal
Purified
RUO
100% homology to SWISS-PROT Q92569, DKFZp686P05226, FLJ41892, p55, Phosphatidylinositol 3-kinase 55 kDa regulatory subunit gamma, phosphoinositide-3-kinase, regulatory subunit 3 (gamma), phosphoinositide-3-kinase, regulatory subunit 3 (p55, gamma), phosphoinositide-3-kinase, regulatory subunit, polypeptide 3 (p55, gamma), PI3K regulatory subunit gamma, PI3-kinase subunit p55-gamma, regulatory subunit, polypeptide 3 (p55, gamma)
PIK3R3
Primary
Western Blot
Unconjugated
Rabbit
Q92569
8503
IgG
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.