Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI 3-Kinase p85 alpha Antibody (13E7), Novus Biologicals™
SDP

Catalog No. NB298213 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298213 100 μL
NB298212 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298213 Supplier Novus Biologicals Supplier No. NBP326382100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PI 3-Kinase p85 alpha Monoclonal antibody specifically detects PI 3-Kinase p85 alpha in Human samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen PI 3-Kinase p85 alpha
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 13E7
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias AGM7, GRB1, IMD36, p85, p85-ALPHA, Phosphatidylinositol 3-kinase 85 kDa regulatory subunit alpha, phosphatidylinositol 3-kinase regulatory subunit alpha, phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha), phosphatidylinositol 3-kinase-associated p-85 alpha, phosphoinositide-3-kinase regulatory subunit alpha, phosphoinositide-3-kinase, regulatory subunit 1 (alpha), PI3K regulatory subunit alpha, PI3-kinase regulatory subunit alpha, PI3-kinase subunit p85-alpha, PIK3R1, PtdIns-3-kinase regulatory subunit alpha, PtdIns-3-kinase regulatory subunit p85-alpha
Host Species Rabbit
Immunogen A synthesized peptide derived from Human PI 3-Kinase p85 alpha [UniProt P27986]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Diabetes Research, Immune Dysfunction, Lipid and Metabolism, mTOR Pathway, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5295
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.