Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI 3-Kinase p85 beta Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB061146 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB061146 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB061146 Supplier Novus Biologicals Supplier No. NBP299487JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PI 3-Kinase p85 beta Polyclonal specifically detects PI 3-Kinase p85 beta in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PI 3-Kinase p85 beta
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias P85B, phosphatidylinositol 3-kinase 85 kDa regulatory subunit beta, phosphatidylinositol 3-kinase regulatory subunit beta, phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 2 (p85 beta), phosphoinositide-3-kinase, regulatory subunit 2 (beta), phosphoinositide-3-kinase, regulatory subunit 2 (p85 beta), phosphoinositide-3-kinase, regulatory subunit, polypeptide 2 (p85 beta), PI3K regulatory subunit beta, PI3-kinase regulatory subunit beta, PI3-kinase subunit p85-beta, ptdIns-3-kinase regulatory subunit beta, ptdIns-3-kinase regulatory subunit p85-beta
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human PI 3-Kinase p85 beta.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5296
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.