Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI 4 Kinase type 2 beta Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB336469 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB336469 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB336469 Supplier Novus Biologicals Supplier No. NBP297902MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PI 4 Kinase type 2 beta Polyclonal antibody specifically detects PI 4 Kinase type 2 beta in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen PI 4 Kinase type 2 beta
Applications Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.1.67, FLJ11105, phosphatidylinositol 4-kinase type 2 beta, phosphatidylinositol 4-kinase type 2-beta, Phosphatidylinositol 4-kinase type II-beta, phosphatidylinositol 4-kinase type-II beta, PI4KIIB, PI4KII-BETA, PIK42B
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PI 4 Kinase type 2 beta fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 55300
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.