Learn More
Invitrogen™ Pi16 Polyclonal Antibody

Description
In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human PI16 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | Pi16 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | PI16 |
| Gene Accession No. | Q9ET66 |
| Gene Alias | 1200009H11Rik; CD364; Cripi; CRISP9; CRISP-9; Cysteine-rich protease inhibitor; cysteine-rich secretory protein 9; microseminoprotein, beta-binding protein; MSMBBP; peptidase inhibitor 16; Pi16; PI-16; protease inhibitor 16; PSEC0164; PSP94-binding protein; PSPBP; UNQ289/PRO328 |
| Gene Symbols | PI16 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.