Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI4K2A Antibody, Novus Biologicals™
SDP

Catalog No. NBP24415805 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP24415805 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP24415805 Supplier Novus Biologicals Supplier No. NBP2441580.05MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PI4K2A Polyclonal specifically detects PI4K2A in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen PI4K2A
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μ/mL, ELISA 1: 10000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 50% (v/v) Glycerol with no preservative
Gene Alias DKFZp761G1923, EC 2.7.1.67, phosphatidylinositol 4-kinase type 2 alpha, phosphatidylinositol 4-kinase type 2-alpha, phosphatidylinositol 4-kinase type II (PI4KII), Phosphatidylinositol 4-kinase type II-alpha, PI4KII, PIK42A, RP11-548K23.6
Gene Symbols PI4K2A
Host Species Rabbit
Immunogen Anti-PI4K2A alpha affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide at the N-terminal of human PI4K2A protein.
Quantity 0.05 mg
Primary or Secondary Primary
Gene ID (Entrez) 55361
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.