Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PI4KB/PI4KIII beta Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB290473 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290473 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290473 Supplier Novus Biologicals Supplier No. NBP212814PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PI4KB/PI4KIII beta Polyclonal antibody specifically detects PI4KB/PI4KIII beta in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen PI4KB/PI4KIII beta
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias DKFZp686O1820, EC 2.7.1, EC 2.7.1.67, FLJ30129, phosphatidylinositol 4-kinase, catalytic, beta, pi4K92, PI4Kbeta, PI4K-beta, PI4KIIIBETA, type III phosphatidylinositol 4-kinase beta, wortmannin-sensitive
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1 and 50 of human Phosphatidylinositol 4-Kinase, Catalytic, beta using the numbering given in entry CAA09495.1 (GeneID 5298).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5298
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.