Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIAS1 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501992136
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2136 25 μL
50-199-2135 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2136 Supplier Rockland Immunochemicals Supplier No. 600401GZ8S
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. A BLAST analysis was used to suggest reactivity to mouse 92.9% homology for the immunogen sequence. Cross-reactivity from PIAS1 family and other sources has not been determined.

PIAS1 functions as an E3-type small ubiquitin-like modifier (SUMO) ligase, stabilizing the interaction between UBE2I and the substrate, and as a SUMO-tethering factor. This protein plays a crucial role in transcriptional coregulation of various cellular pathways, including the STAT pathway, the p53 pathway and the steroid hormone signaling pathway. It functions in testis as a nuclear receptor transcriptional coregulator and may have a role in androgen receptor initiation and maintenance of spermatogenesis. The effects of transcriptional coregulation, transactivation or silencing, may vary depending upon the biological context.

Specifications

Antigen PIAS1
Applications ELISA, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.01M sodium phosphate with 0.25M NaCl and 0.01% sodium azide; pH 7.2
Gene PIAS1
Gene Accession No. O75925
Gene Alias 2900068C24Rik; AR interacting protein; Ddxbp1; DEAD/H (Asp-Glu-Ala-Asp/His) box binding protein 1; DEAD/H box-binding protein 1; E3 SUMO-protein ligase PIAS1; E3 SUMO-protein transferase PIAS1; GBP; Gu binding protein; GU/RH-II; Gu-binding protein; PIAS1; protein inhibitor of activated STAT 1; Protein inhibitor of activated STAT protein 1; protein inhibitor of activated STAT, 1; protein inhibitor of activated STAT-1; RING-type E3 ubiquitin transferase PIAS1; RNA helicase II-binding protein; zinc finger, MIZ-type containing 3; ZMIZ3
Gene Symbols PIAS1
Host Species Rabbit
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8554
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.