Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIAS2 Antibody (OTI2B5), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB301970 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301970 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301970 Supplier Novus Biologicals Supplier No. NBP273398APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PIAS2 Monoclonal antibody specifically detects PIAS2 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen PIAS2
Applications Western Blot
Classification Monoclonal
Clone OTI2B5
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias Androgen receptor-interacting protein 3, ARIP3, DAB2-interacting protein, DIP, E3 SUMO-protein ligase PIAS2, MGC102682, miz, Miz1, Msx-interacting zinc finger protein, Msx-interacting-zinc finger, PIAS-NY protein, PIASX, PIASX-ALPHA, PIASX-BETA, Protein inhibitor of activated STAT x, protein inhibitor of activated STAT, 2, Protein inhibitor of activated STAT2, SIZ2, zinc finger, MIZ-type containing 4, ZMIZ4
Host Species Mouse
Immunogen Full length human recombinant protein of human PIAS2(NP_004662) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 9063
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.