Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ovalbumin Antibody, Novus Biologicals™
SDP

Catalog No. NB600922SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600922SS 0.02 mg
NB60092201 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB600922SS Supplier Novus Biologicals Supplier No. NB600922SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 7 publications

Ovalbumin Polyclonal specifically detects Ovalbumin in Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Single Cell Western.

Specifications

Antigen Ovalbumin
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:5000-1:20000, Immunohistochemistry, Immunoprecipitation, Single Cell Western 1:10
Formulation Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. P01012
Gene Alias Allergen Gal d 2, CAP 2, CAP2, Cytoplasmic antiproteinase 2, Egg albumin, Gal d II, Hen egg white, PI8, Plakalbumin, Protease inhibitor 8, Serpin B8, serpin peptidase inhibitor clade B member 8, SERPINB14, SERPINB8
Gene Symbols OVAL
Host Species Rabbit
Immunogen Ovalbumin [Hen Egg White] (Uniprot: P01012)
Molecular Weight of Antigen 42.88 kDa
Purification Method Multi-step
Quantity 0.02 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 396058
Test Specificity Cross reactivity against Ovalbumin from other sources is unknown.
Reconstitution For 0.1 mg size: Reconstitute with 10 ul deionized water (or equivalent) to a concentration of 10 mg/mL. The 0.02 mg size is in liquid format.
Target Species Chicken, Virus
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.