Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ovalbumin Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 7 publications

$254.00 - $670.00

Specifications

Antigen Ovalbumin
Classification Polyclonal
Conjugate Unconjugated
Form Antisera
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB600922SS
SDP
View Documents
Novus Biologicals
NB600922SS
0.02 mg
Each for $254.00
Only null left
Add to Cart
 
NB60092201
SDP
View Documents
Novus Biologicals
NB6009220.1MG
0.1 mg
Each for $670.00
Only null left
Add to Cart
 
Description

Description

Ovalbumin Polyclonal specifically detects Ovalbumin in Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Single Cell Western.
Specifications

Specifications

Ovalbumin
Unconjugated
Rabbit
Chicken, Virus
Allergen Gal d 2, CAP 2, CAP2, Cytoplasmic antiproteinase 2, Egg albumin, Gal d II, Hen egg white, PI8, Plakalbumin, Protease inhibitor 8, Serpin B8, serpin peptidase inhibitor clade B member 8, SERPINB14, SERPINB8
IgG
Polyclonal
Antisera
RUO
P01012
396058
Primary
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.