Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piccolo Antibody (4G3.3), Novus Biologicals™
SDP

Catalog No. NB019451 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019451 0.025 mg
NBP229749 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019451 Supplier Novus Biologicals Supplier No. NBP2297490.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Piccolo Monoclonal specifically detects Piccolo in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Piccolo
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4G3.3
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q9Y6V0
Gene Alias piccolo (presynaptic cytomatrix protein), protein piccolo
Gene Symbols PCLO
Host Species Mouse
Immunogen GST-tagged recombinant protein.
Molecular Weight of Antigen 547 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27445
Test Specificity This antibody recognizes piccolo.
Target Species Mouse, Rat
Content And Storage Store at 4C. Do not freeze.
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.