Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Piccolo Antibody (4G3.3), Novus Biologicals™

Mouse Monoclonal Antibody
$214.28 - $689.94
Specifications
| Antigen | Piccolo |
|---|---|
| Clone | 4G3.3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
Description
Piccolo Monoclonal specifically detects Piccolo in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Piccolo | |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| RUO | |
| Q9Y6V0 | |
| 27445 | |
| GST-tagged recombinant protein. | |
| Primary | |
| Store at 4C. Do not freeze. | |
| 547 kDa |
| 4G3.3 | |
| Western Blot 1:100-1:2000, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Mouse | |
| Mouse, Rat | |
| piccolo (presynaptic cytomatrix protein), protein piccolo | |
| PCLO | |
| IgM κ | |
| Affinity Purified | |
| This antibody recognizes piccolo. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title