Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piccolo Antibody (PCLO-01), FITC, Novus Biologicals™
SDP

Catalog No. NB002479 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002479 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002479 Supplier Novus Biologicals Supplier No. NBP262209F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Piccolo Monoclonal specifically detects Piccolo in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Piccolo
Applications Flow Cytometry
Classification Monoclonal
Clone PCLO-01
Conjugate FITC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias piccolo (presynaptic cytomatrix protein), protein piccolo
Gene Symbols PCLO
Host Species Mouse
Immunogen Human recombinant PCLO protein
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 27445
Test Specificity The antibody A53-B/A2 reacts with Rod domain of cytokeratin 19 (40 kDa) in human tissue. Cytokeratin 19 is not expressed in hepatocytes; it is often co-expressed with cytokeratin 7.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.