Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Picus™ NxT Electronic Pipette, Single Channel, Trade-In

Catalog No. 14557824PR Shop All Sartorius Products
Change view
Click to view available options
Volume (Metric):
0.2 to 10 μL
5 to 120 μL
50 to 1000 μL
Color:
Blue
Gray
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
14-557-824PR 50 to 1000 μL Blue
14-557-821PR 0.2 to 10 μL Gray
145-578-22PR 5 to 120 μL Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-557-824PR Supplier Sartorius Supplier No. LH745081PR

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Picus™ NxT Electronic Pipette, Single Channel is designed to ease user's workload and to protect from repetitive strain injury (RSI).

Laboratory work requires repetition of similar pipetting steps that may cause strain on hand and arm, and thus may compromise results. To minimize this risk, a highly ergonomic electronic pipette should be used for minimum effort and strain. Sartorius Picus™ NxT are the most sophisticated and ergonomic electronic compact and light weight pipettes.

  • Password protection for stored programs
  • Very last liquid droplets dispensed by repeated blow-out for maximum recovery
  • Features one-touch operating-button and feather-light electronic tip ejection
  • Includes plate tracker for microwell plate dispensing
  • Electronic piston control and brake for precise pipetting
  • Safe-cone filters to prevent contamination
  • Adjustment wheel for volume setting and navigation
  • Range of pipetting modes for diluting and titrating to serial dispensing
  • Intuitive user interface in a choice of language options: English, French, German, Russian and Chinese
  • Certificate of accredited 3-point calibration
  • RFID-enabled to support asset management

Applications: PCR and other DNA/RNA techniques, ELISA, Protein analysis and Cell culture

Offer Details
After purchasing this trade-in program item please follow these steps to complete your trade in:
1. Complete Sartorius redemption form. (Contact Fisher Customer Service to obtain the form.)
2. Sartorius will email a link to a prepaid UPS shipping label, shipping instructions, and decontamination form.
3. Visit UPS site, print the shipping label, pack pipette, and ship to Sartorius.

Note: Any brand pipette, in any condition can be traded in. Pipettes must be free of contamination and returned together with the signed Decontamination Form.

TRUSTED_SUSTAINABILITY

Specifications

Calibration Points Single
Certifications/Compliance CE Marked, Certificate of accredited 3-point calibration
Compatible Tips Universal Design, Safetyspace, Optifit
Autoclavable Not Autoclavable
Color Blue
Disposable Reusable
Number of Channels 1
Product Line Picus
Type Electronic Pipette
Volume (Metric) 50 to 1000 μL
Warranty 2 Year
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.