Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pierce-1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017292 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017292 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017292 Supplier Novus Biologicals Supplier No. NBP281981
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Pierce-1 Polyclonal specifically detects Pierce-1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunocytochemistry.

Specifications

Antigen Pierce-1
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, ELISA, Immunocytochemistry/Immunofluorescence 2.5 ug/mL, Immunocytochemistry 2.5 ug/ml
Formulation PBS
Gene Alias C9orf116, chromosome 9 open reading frame 116, FLJ13945, hypothetical protein LOC138162, MGC29761, Pierce 1, RbEST47, RP11-426A6.4
Gene Symbols C9ORF116
Host Species Rabbit
Immunogen Pierce-1 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human Pierce-1. The immunogen is located within the first 50 amino acids of Pierce-1. Amino Acid Squence: APKATAPPERTSDYYR
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 138162
Test Specificity Pierce-1 antibody is predicted to not cross-react with other UPF0691 family members.
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.