Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 1-Step™ ABTS Substrate Solution

Catalog No. pi37615
Click to view available options
Quantity:
250 mL

Ready-to-use, water-soluble peroxidase substrate tablets that yield a measurable green end-product for use in ELISA methods.

Thermo Scientific™ Pierce ABTS Substrates are tablets and a ready-to-use solution of a water-soluble peroxidase substrate that yields a measurable green end product for use in ELISA methods.

ABTS (2,2'-Azinobis [3-ethylbenzothiazoline-6-sulfonic acid]-diammonium salt) is a water-soluble HRP substrate that yields a green end product upon reaction with peroxidase. The green product has two major absorbance peaks, 410nm and 650nm. ABTS is less sensitive than OPD and TMB in ELISA applications. It is less readily oxidized, and its color development is slower (approximately 20 minutes). This may be advantageous if unacceptable background results from the use of the OPD or TMB substrates due to higher sensitivities.

Highlights:

  • Excellent choice when maximum sensitivities are not required
  • Less readily oxidized and color development is slower (approximately 20 minutes) than OPD and TMB
TRUSTED_SUSTAINABILITY

Specifications

Description Pierce 1-Step ABTS Solution
Product Type ELISA Substrate
Quantity 250 mL
Detection Method Colorimetric
Product Line 1-Step™
Substrate Properties Chemical Substrate
Substrate Type HRP (Horseradish Peroxidase)
Content And Storage Store at 4°C

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.