Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Acetonitrile (ACN), LC-MS Grade

Catalog No. pi85188
Click to view available options
Quantity:
1 L
4 x 1 L

An LC/MS-grade preparation with high purity and low UV-absorptivity that makes it suitable for HPLC and mass spectrometry applications.

Thermo Scientific™ Acetonitrile (ACN) is an LC/MS-grade preparation with high purity and low UV-absorptivity that makes it suitable for HPLC and mass spectrometry applications.

Thermo Scientific LC/MS Grade Acetonitrile is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance to provide you with the most sensitive detection across all wavelengths. Acetonitrile is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen atmosphere with PTFE-lined fluorocarbon caps for ultimate protection.

Highlights:

  • 37 quality tests ensure low, stable baselines and lot-to-lot consistency
  • Low UV absorbance yields low baselines and high detection sensitivity
  • Low impurities protects columns and simplifies analysis by eliminating extraneous peaks
  • Specially purified and tested to the highest specifications to ensure data integrity, maximize assay sensitivity, and to prolong equipment life
  • Store between 2°C to 25°C in a dry, cool, well-ventilated area, away from incompatible materials

Product Details:

  • Alternative names: Methyl cyanide, cyanomethane, ethanenitrile
  • Molecular formula: CH3CN
  • Molecular weight: 41.05
  • Density: 0.780 g/mL
TRUSTED_SUSTAINABILITY

Specifications

Grade LC-MS
Item Name Acetonitrile (ACN)
Quantity 4 x 1 L
Content And Storage Store between 2 to 25°C in a dry, cool, well-ventilated area, away from incompatible materials.
Form Liquid
Reagent Type Solvent (Pure)
Product Line Pierce™
For Use With HPLC, Mass Spectrometry
CAS 75-05-8

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.