Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BMOE (bismaleimidoethane)

Catalog No. pi22323
Click to view available options
Quantity:
50 mg

Crosslink sulfhydryls to sulfhydryls (cysteine residues in proteins or peptides) with this short crosslinker composed of maleimide groups and a 7-atom (8.0A) spacer arm.

Thermo Scientific™ Pierce BMOE is a short-arm, maleimide crosslinker for covalent, irreversible conjugation between sulfhydryl groups (e.g., protein or peptide cysteines).

Highlights:

  • Reactive groups: maleimide (homobifunctional)
  • Reactive toward: sulfhydryl groups
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Sulfhydryl-Sulfhydryl
Cleavable No
Description BMOE
Molecular Weight (g/mol) 220.18
PEGylated No
Spacer Arm Length 8.0 Å
Content And Storage Upon receipt store at 4°C.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety Maleimide
Spacer Short (<10 Å)
Form Solid
Quantity 50 mg
Solubility DMF, DMSO
Format Standard
Water Soluble No
Product Type Crosslinker
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.