Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Chloronaphthol Substrate Powder (4-chloro-1-naphthol)

Catalog No. pi34010
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Powders, tablets and ready-made 4-chloro-1-naphthol solutions for chromogenic HRP detection in Western blot and tissue staining methods.

Thermo Scientific™ Pierce Chloronaphthol (4-CN) Substrates are powders, tablets and ready-made solutions of 4-chloro-1-naphthol for chromogenic detection of horseradish peroxidase (HRP) activity in blotting and tissue staining methods.

Chloronaphthol (4-CN, MW = 178.6) reacts with HRP to form a distinct blue to blue-purple product to localize and visualize the activity of peroxidase-conjugated antibodies on blots or fixed tissue samples. The HRP substrate is not as sensitive or stable as other chromogenic substrates, such as TMB and DAB. However, the blue-purple precipitate photographs well, and its alcohol-solubility is a property that can be utilized in certain double-staining applications.

Highlights:

  • HRP substrate – for detection of horseradish peroxidase activity on solid media

  • Chromogenic – no special equipment needed for visualization

  • Blue-purple – easy to photograph in color or black-and-white

  • Package options – choose powder, tablets, or ready-to-use solution

TRUSTED_SUSTAINABILITY

Specifications

Product Type Substrate
Detection Method Colorimetric
Form Powder
Quantity 25 g
Substrate Horseradish Peroxidase
Content And Storage Store at 4°C.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.