Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Dimethylformamide (DMF), Sequencing grade PROMO

Catalog No. PI20673PM
Click to view available options
Quantity:
50 mL

Catalog No. PI20673PM

Supplier: Thermo Scientific™ 20673

Perform liquid chromatography (HPLC) and mass spectrometry (LC-MS) applications with this 99.5%-pure DMF solvent (MW 73.09, CAS 68-12-2) with low UV-absorptivity.

This sequencing-grade dimethylformamide is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance to provide you with the most sensitive detection across all wavelengths. DMF is packaged in solvent-rinsed amber glass bottles and sealed with PTFE-lined fluorocarbon caps for ultimate protection.

Highlights:

  • Highly purified and tested to ensure reliable performance in HPLC, mass spectrometry and sequencing applications
  • Packaged in solvent-rinsed amber glass bottles and sealed with PTFE-lined fluorocarbon caps

Product Details:

  • General properties of DMF
  • Alternative names: n,n-Dimethylformamide, n-Formyldimethylamine
  • Molecular formula: C3H7ON
  • Molecular weight: 73.09
  • Density:0.945g/mL
  • CAS number: 68-12-2

Pierce sequencing-grade DMF is specially purified and tested to the highest specifications to ensure the integrity of your data, maximize sensitivity in your assay and prolong the life of your equipment.

Specifications

Item Name Dimethylformamide (DMF)
Quantity 50 mL
Content And Storage Store at room temperature.
Form Liquid
Reagent Type Solvent (Pure)
Product Line Pierce
For Use With HPLC, Mass Spectrometry

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.