Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Dimethylsulfoxide (DMSO), LC-MS Grade PROMO

Catalog No. PI85190PM
Click to view available options
:
Thermo Scientific™ Pierce™ Dimethylsulfoxide (DMSO), LC-MS Grade

Catalog No. PI85190PM

Supplier: Thermo Scientific™ 85190

Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the Fisher Scientific channel.

A sequencing-grade preparation with high purity and consistent absorptivity properties, for use in HPLC and mass spectrometry methods.

A sequencing-grade preparation with high purity for use in traditional amino acid analyzers and other high-volume applications.

This sequencing-grade dimethylsulfoxide is >99.5% pure DMSO. The economically priced 950 mL bottles are an excellent choice for protein and peptide applications that requiring large volumes of the water-miscible solvent. The 50 mL bottles are provide convenience and minimize waste for small dissolution and reaction volumes. The purity test ensures that preparation is essentially devoid of amines. The formulation was originally developed for peptide sequencing applications with amino acid analyzers, which required high purity solvent and an affordable price. This DMSO is packaged in solvent-rinsed amber glass bottles and sealed with PTFE-lined fluorocarbon caps for ultimate protection.

Highlights:

  • High-purity (low amine) solvent for various protein, peptide and amino acid applications
  • Packaged in solvent-rinsed amber glass bottles and sealed with PTFE-lined fluorocarbon caps
  • Suitable for HPLC detection, although it is not specifically tested for low UV-absorptivity

Product Details:

  • General properties of DMSO
  • Alternative names: Methylsulfinylmethane, methyl sulfoxide
  • Molecular formula: C2H6SO
  • Molecular weight: 78.13
  • Density: 1.101 g/mL
  • CAS number: 67-68-5

Pierce sequencing-grade DMSO is specially purified and tested to the highest specifications to ensure the integrity of your data, maximize sensitivity in your assay and to prolong the life of your equipment.

Specifications

Grade LC-MS
Item Name Dimethylsulfoxide (DMSO)
Quantity 50 mL
Content And Storage Store at room temperature.
Form Liquid
Reagent Type Solvent (Pure)
Product Line Pierce™
For Use With Liquid Chromatography, Mass Spectrometry
Packaging Amber Glass Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.