Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ ECL Plus Western Blotting Substrate

Catalog No. p-9349003
Change view
Click to view available options
Quantity:
100 mL Kit
300 mL Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FER32132X3 300 mL Kit
FER32132 100 mL Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FER32132X3 Supplier Thermo Scientific™ Supplier No. 32132X3
Only null left
Add to Cart
Add to Cart

Acridan-based chemiluminescent and chemifluorescent HRP substrate for Western blot detection using X-ray film or CCD- or laser-based imagers.

Thermo Scientific™ Pierce ECL Plus Substrate is an acridan-based chemiluminescent and chemifluorescent substrate to detect peroxidase activity from HRP-conjugated probes by Western blotting using X-ray film, CCD or laser-based imagers.

Thermo Scientific Pierce ECL Plus Substrate is an acridan-based chemiluminescent and chemifluorescent HRP substrate for western blot detection using X-ray film or CCD- or laser-based imagers. This product is sold as Pierce ECL 2 Substrate (Cat. #PI80196X3) through Fisher Scientific and other channels.

Features of ECL Plus Substrate:

  • Easy to use—can be substituted for the discontinued GE ECL Plus Substrate without any re-optimization
  • Higher sensitivity—detect targets down to the low-picogram level
  • Longer signal duration—sustained light output for as long as 5 hours
  • More imaging options—X-ray, CCD or laser-based imagers
  • More affordable—high quality and performance at a lower price than other suppliers' similar substrates

Pierce ECL Plus Substrate generates acridinium esters when it reacts with HRP. As these ester intermediates react with peroxide, they produce a strong and sustained chemiluminescent signal that can be captured by X-ray film CCD imaging. The substrate also produces a robust fluorescent signal that can be captured with fluorescence imagers. Pierce ECL Plus Substrate enables the detection of low-picogram amounts of target protein on nitrocellulose or PVDF membrane when probed with appropriate primary and secondary antibody concentrations. Its broad dynamic range, high sensitivity and long-lasting signal make Pierce ECL Plus Substrate an excellent choice for western blot analysis.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Substrate
Detection Method Chemiluminescence, Fluorescence
Form Liquid
For Use With (Application) Western Blot
Membrane Compatibility Nitrocellulose, PVDF
Product Line Pierce
Quantity 300 mL Kit
Sensitivity Low picogram to mid-femtogram
Sufficient For 3000 cm2 of membrane
Kit Contents Detection Reagent A, 3 x 100 mL, Detection Reagent B, 3 x 2.5 mL
Content And Storage Upon receipt store at 4°C. Product shipped on ice.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.