Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Pierce™ Endotoxin-Free Magnesium Chloride (MgCl2) (10 mM) PROMO
Click to view available options
:
Thermo Scientific™ Pierce™ Endotoxin-Free Magnesium Chloride
Description
Pierce Endotoxin-Free MgCl2 is suitable for the preparation of samples for endotoxin detection (LAL) assay. As a diluent, it will eliminate the inhibitory chelation effects of EDTA. It contains <0.005 EU/mL endotoxin.
Specifications
Specifications
Concentration | 10 mM |
For Use With (Equipment) | LAL Endotoxin Assay |
Quantity | 5 x 10 mL |
Packaging | Vial |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction