Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Fast Blocking Buffer

Catalog No. p-4589209
Change view
Click to view available options
Quantity:
100 mL
500 mL
Catalog No. Quantity
PI37575 500 mL
PI37576 100 mL
2 options

Catalog No. PI37575

Supplier: Thermo Scientific™ 37575

Only null left
Add to Cart
Edge
Add to Cart

Optimize Western blotting systems by decreasing both background signal and incubation time compared to traditional blocking agents with this ready-to-use blocker solution.

Optimize Western blotting systems by decreasing both background signal and incubation time compared to traditional blocking agents with Thermo Scientific™ Pierce Fast Blocking Buffer, a ready-to-use blocker solution.

Additional Information and Online Ordering

Pierce Fast Blocking Buffer streamlines Western blot protocols by providing consistent, high-quality blocking for nitrocellulose or PVDF membranes in only 5 minutes. Blocking and probing steps with Fast Blocking Buffer prevents nonspecific binding of detection antibodies during probing steps but allows specific detection to occur. Pierce Fast Wash Buffer provides efficient membrane washing after incubation with a primary and secondary antibody to effectively shorten total washing time to 20 minutes (from the typical 90 minutes). These two buffers are compatible with antibodies and biotin-avidin systems, as well as both nitrocellulose and PVDF membranes. The blocking buffer provides rapid and effective blocking in just five minutes while producing results comparable to traditional blockers for Western blots.

Highlights:

  • Fast – use both buffers to shorten the typical Western blot development by over 2 hours

  • Simple – optimized protocol makes Western blot analysis easier than ever

  • Low background – provides results comparable to classic Western blotting buffers

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Blocking Buffer
Concentration 1X
For Use With (Application) Western Blot
Physical Form Liquid
Quantity 500 mL
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.