Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Pierce™ Heptafluorobutyric Acid (HFBA), HPLC Grade PROMO
Click to view available options
Quantity:
10 x 1 mL
Description
Highlights:
- Typical purity is 99.7% by GC; <0.1% water
- Sequencing reagent for classical and automated Edman degradation of peptides and proteins
- Density: 1.645
- B.P. 120°C
- Packaged under nitrogen in amber glass ampules or bottles
- Clear, colorless liquid
Ordering Information:
X in the ordering table indicates that hazardous shipping charges apply.
Specifications
Specifications
| Item Name | Heptafluorobutyric Acid (HFBA) |
| Content And Storage | Store at room temperature. |
| Form | Liquid |
| Reagent Type | Ion Pairing Reagent |
| Quantity | 10 x 1 mL |
| Formulation | 99.5% HFBA |
| Product Line | Pierce |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction