Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Pierce™ Heptafluorobutyric Acid (HFBA), Sequencing Grade PROMO
Click to view available options
Quantity:
100 mL
Description
Highlights:
- Typical purity is 99.7% by GC; <0.1% water
- Sequencing reagent for classical and automated Edman degradation of peptides and proteins
- Density: 1.645
- B.P. 120°C
- Packaged under nitrogen in amber glass ampules or bottles
- Clear, colorless liquid
Ordering Information:
X in the ordering table indicates that hazardous shipping charges apply.
Specifications
Specifications
| Item Name | Heptafluorobutyric Acid (HFBA) |
| Content And Storage | Store at room temperature. |
| Form | Liquid |
| Reagent Type | Ion Pairing Reagent |
| Quantity | 100 mL |
| Formulation | 99.5% HFBA |
| Product Line | Pierce |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction