Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Lane Marker Reducing Sample Buffer
GSA_VA

Catalog No. pi39000
Click to view available options
Quantity:
5 mL

Ready-to-use 5X SDS-PAGE sample loading buffers available in reducing and nonreducing formulations, both with a pink dye buffer-front marker.

Thermo Scientific™ Pierce Lane Marker Sample Buffers are ready-to-use 5X SDS-PAGE sample loading buffers, available in reducing and nonreducing formulations, with a pink dye buffer-front marker.

Lane Marker Reducing and Non-Reducing Sample Buffers are convenient and ready to use for SDS-polyacrylamide gel electrophoresis (SDS-PAGE) and Western blotting. Formulated as 5X stocks rather than the traditional 2X stocks, they enable a larger volume of protein solution to be included in the sample that is loaded in each well. Particularly unique in these buffers is the use of a bright pink tracking dye instead of the traditional bromophenol blue dye. This pink dye marks not only the dye front in the gel but also transfers and permanently binds to nitrocellulose membranes. Consequently, the dye can be used to assess transfer efficiency and align cut membranes. The Reducing Sample Buffer contains DTT as the reducing agent, eliminating the repugnant odor associated with mercaptoethanol-containing buffers.

Lane Marker Sample Buffers are easy to use. Simply mix one part Sample Buffer with four parts protein sample, heat denature, and then load the gel sample wells. Visualize progress of the electrophoresis run by monitoring the location of the pink dye front. Lane Marker Sample Buffers may be used in denaturing gels of all kinds and are compatible with coomassie dye and silver staining techniques, as well as Western blotting procedures.

Highlights:

  • Easy to see – bright pink dye marks the buffer front, clearly indicating the progress of the electrophoresis run

  • Concentrated – 5X stocks enable a greater range of protein sample volumes to be used for electrophoresis

  • Transferable marker dye – pink dye transfers with protein to nitrocellulose membrane, enabling verification of transfer direction and orientation of the blot

  • Flexibility in SDS-PAGE applications – available in two ready-to-use formulations for reducing or non-reducing denaturing SDS polyacrylamide electrophoresis

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store single-use aliquots at -5 to -30°C.
Description Pierce Lane Marker Reducing Sample Buffer (5X)
Buffer Sample Loading Buffers
Concentration 5X
Detection Method Fluorescence
Gel Type SDS-PAGE
Product Type Lane Marker Reducing Sample Buffer
Gel Compatibility SDS-PAGE Gels
Product Line Pierce™
Quantity 5 mL
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.