Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ OPD Substrate Tablets (o-phenylenediamine dihydrochloride)
Click to view available options
Quantity:
50 tablets
Description
Thermo Scientific™ Pierce OPD is a water-soluble substrate for horseradish peroxidase (HRP) that produces a yellow-orange product detectable at 492nm by ELISA plate readers.
OPD (o-phenylenediamine dihydrochloride) yields a water-soluble yellow-orange product when reacted with peroxidase with an absorbance maximum of 492nm. OPD can easily be dissolved in a substrate buffer such as Thermo Scientific Stable Peroxide Substrate Buffer (Part No. 34062).
Highlights:
- Water soluble product when reacted with peroxidase
- Absorbance maximum of 492nm

Specifications
Specifications
Description | Pierce OPD Tablets |
Product Type | ELISA Substrate |
Quantity | 50 tablets |
Detection Method | Colorimetric |
Substrate Properties | Chemical Substrate |
Substrate Type | HRP (Horseradish Peroxidase) |
Content And Storage | Upon receipt store product at 4°C. Product is shipped at ambient temperature. |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction