Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Premium Grade Sulfo-NHS (N-hydroxysulfosuccinimide)

Catalog No. p-7000097
Click to view available options
Quantity:
500 mg
10 g

Optimize applications where product integrity and risk minimization are paramount with our highest quality formulation of this N-hydroxysuccinimide modification reagent.

Optimize applications where product integrity and risk minimization are paramount with the specially characterized Thermo Scientific™ Pierce™ Premium Grade Sulfo-NHS, our highest quality formulation of this N-hydroxysuccinimide chemical modification reagent.

Thermo Scientific Pierce Premium Grade Sulfo-NHS is our highest quality formulation of this N-hydroxysuccinimide chemical modification reagent, specially characterized for applications where product integrity and risk minimization are paramount.

Compared to the standard grade product, Premium Grade Sulfo-NHS provides more clearly defined quality and product support by including (a) increased analytical testing and product characterization, (b) greater batch-specific information and quality assurance review, (c) extensive lot sample retention and (d) change control notification. Sulfo-NHS (N-hydroxysulfosuccinimide) enables control and modification of carbodiimide crosslinking reactions involving activation of carboxylates (–COOH) for conjugation with primary amines (–NH2). Derivatives are easily synthesized by mixing the sulfo-NHS with a carboxyl-containing molecule and a dehydrating agent such as the carbodiimide EDC (EDAC). The method is the basis for generating many types of protein labeling reagents, including amine-reactive fluorescent dyes, biotin affinity tags and pegylation compounds.

Highlights:
  • Sulfo-NHS – use with EDC crosslinker to prepare amine-reactive NHS ester reagents
  • High quality – identity and purity confirmed by several tests, including quantitative NMR
  • Product integrity – enhanced level of testing and characterization compared to standard grade
  • Lot retention – ample supply of past lots retained to ensure future process testing
  • Change management – Change Control Notification (CCN) service
  • Consistent manufacture – batch-specific manufacturing documentation review
Applications:
  • Improve efficiency of EDC coupling reactions
  • Convert carboxyls to amine-reactive sulfo-NHS esters
  • Membrane-impermable for cell surface labeling
  • Crosslink proteins to carboxyl-coated beads or surfaces more efficiently
  • Activate nanoparticles with amine-reactive sulfo-NHS esters
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Carboxyl
Cleavable No
Molecular Weight (g/mol) 217.13
PEGylated No
Content And Storage Upon receipt store at 4°C.
Cell Permeability No
Shipping Condition Ambient
Product Line Pierce™
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Carbodiimide
Spacer Short (<10 Å)
Form Powder
Quantity 10 g
Solubility Water
Format Standard, Single-use, Premium-grade
Water Soluble Yes
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.