Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Protein-Free Blocking Buffer

Catalog No. p-7184558
Click to view available options
Quantity:
100 mL
1 L
Chemical Name or Material:
Protein-Free (PBS)
Protein-Free (TBS)
Protein-Free T20 (PBS)
Protein-Free T20 (TBS)

Pierce Protein-Free Blocking Buffer is a non-protein compound that provides effective blocking for membrane-based and plate-based protein detection methods, resulting in extremely low background.

Pierce Protein-Free Blocking Buffer is a non-protein compound that provides effective blocking for membrane-based and plate-based protein detection methods, resulting in extremely low background. It is supplied in a convenient, ready-to-use format.

Features of Protein-Free Blocking Buffer include:
Protein-free blocker—minimizes or eliminates cross-reactivity associated with protein-based blocking buffers
Applications—compatible with western blotting (membranes), ELISA (microplates), and arrays (coated glass slides)
Streptavidin-friendly—free of biotin; no interference with avidin-biotin detection systems
High-performance—optimized and validated in many protein methods to provide high signal-to-noise ratio (i.e., no quenching of specific binding and signal but eliminating nonspecific binding and background)

Traditional blocking buffers contain proteins that can cross-react in certain immunodetection systems, resulting in high background and reduced signal. Pierce Protein-Free Blocking Buffers are devoid of protein while remaining highly effective at blocking plates, membranes, and other surfaces for ELISA, western blotting, glass slide arrays, and other applications.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Protein-Free (PBS)
Concentration 1X
For Use With (Application) ELISA
Physical Form Liquid
Quantity 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.