Learn More
Thermo Scientific™ Pierce™ Pyrogen-Free Glass Test Tubes PROMO
Click to view available options
:
Thermo Scientific™ Pierce™ Pyrogen-Free Glass Test Tubes
Description
These endotoxin-free glass test tubes contain less than 0.005 EU/mL endotoxin. They are recommended for use as reaction tubes in endotoxin detection assays.
Specifications
Specifications
| Material | Glass |
| For Use With | Gel Clot and Chromogenic Endotoxin Assay |
| Quantity | 30 Tubes |
| Product Type | Test Tube |
For Research Use Only. Not for use in diagnostic procedures.