Learn More
Thermo Scientific™ Pierce™ Recombinant Protein A/G, Peroxidase Conjugated PROMO
Description
The extended Fc-binding properties of Protein A/G make it a popular tool in the investigation and purification of immunoglobulins. Protein A/G binds to all human IgG subclasses, IgA, IgE, IgM and to a lesser extent IgD; however, it does not bind mouse IgA, IgM or murine serum albumin. Protein A/G is an excellent tool for purification and detection of mouse monoclonal antibodies from IgG subclasses without interference from these other serum proteins. Individual subclasses of mouse monoclonals are most likely to have stronger affinity to this chimeric protein than to either Protein A or Protein G.
Highlights:
- Contains four Fc-binding domains from Protein A and two from Protein G
- Provides binding for all antibody species and subclasses recognized by either Protein A or Protein G
Specifications
Specifications
Product Type | Protein A/G, Peroxidase Conjugate |
Conjugate | HRP |
Quantity | 0.5 mg |
Product Line | Pierce™ |
Form | Powder |
Protein Subtype | Protein A |
Target | Antibodies |
Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C. |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.