Learn More
Thermo Scientific™ Pierce™ Recombinant Protein G, 5 mg PROMO
Description
Protein G is a bacterial cell wall protein isolated from group G Streptococci. DNA sequencing of native Protein G identifies two IgG-binding domains and sites for albumin and cell surface binding. The albumin and cell surface binding domains have been eliminated from Recombinant Protein G to reduce nonspecific binding and, therefore, can be used to separate IgG from crude samples. Optimal binding occurs at pH 5, although binding is also effective at pH 7.0 to 7.2.
Highlights:
- Contains two Fc-binding domains per protein
- Better than Protein A for mouse (incl. IgG1), human (incl. IgG3) and rat, goat and cow antibodies
- Poorer than Protein A for guinea pig, pig, dog and cat
- Does not bind human IgM, IgD or IgA
Specifications
Specifications
Product Type | Protein G |
Quantity | 5 mg |
Product Line | Pierce™ |
Form | Powder |
Protein Subtype | Protein G |
Target | Antibodies |
Content And Storage | Upon receipt store at -20°C. Product is shipped at ambient temperature. |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.