Learn More
Thermo Scientific™ Pierce™ S-Nitrosylation Western Blot Kit, 40 Reactions PROMO
Description
S-nitrosylation is a post-translational modification that regulates numerous processes, including cellular proliferation, apoptosis, smooth muscle relaxation, neurotransmitter release, and differentiation. During S-nitrosylation, nitric oxide radicals react with cysteine thiols to produce an S-NO adduct that alters protein activity.
Pierce S-Nitrosylation Western Blot Kit
The Pierce S-Nitrosylation Western Blot Kit provides the necessary reagents to perform a modified version of the traditional S-nitrosylation switch assay, including a cysteine-blocking reagent, reducing agent, reaction buffer, iodoTMT labeling reagent, and anti-TMT antibody. Labeling is accomplished using the non-biological iodoTMT reagent instead of HPDP-biotin, reducing background signal during western blot detection. In addition to detection of S-nitrosylated proteins by western blot, an immobilized anti-TMT resin can be used to selectively enrich S-nitrosylated proteins/peptides labeled with iodoTMT reagents. This workflow allows for S-nitrosylation site mapping and multiplexed quantitation using mass spectrometry.
Features of Pierce S-Nitrosylation Western Blot Kit include:
- Specific—optimized labeling reactions switch S-nitrosocysteine post translational modifications with iodoTMT reagent for easy detection by western blot using anti-TMT antibody
- Better signal-to-noise—uses a non-biological iodoTMT reagent for labeling instead of HPDP-biotin, resulting in less background during western blot detection
- Workflow-compatible—labeling method is compatible with alternative methods of analysis, including enrichment via anti-TMT resin and multiplex quantitation by mass spectrometry
- Convenient—all needed reagents included one kit
HENS Buffer
HENS Buffer consists of 100 mM HEPES (pH 7.8), 1 mM EDTA, 0.1 mM Neocuproine, and 1% SDS. It is a cell lysis buffer intended for use with the Pierce S-Nitrosylation Western Blot Kit, which enables sensitive detection of protein S-nitrosocysteine post-translational modifications in a complete, easy-to-use kit.
Specifications
Specifications
Detection Method | S-Nitrosylation |
Product Line | Pierce™ |
Quantity | 40 Reactions |
Reactivity | All species |
Content And Storage | Upon receipt store Kit at –20°C, or store individual components as indicated above. Kit is shipped with an ice pack. |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.