Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Sulfo-NHS (N-hydroxysulfosuccinimide), No-Weigh™ Format
Click to view available options
Quantity:
10 x 2 mg
500 mg
5 g
Description
Thermo Scientific™ Pierce Sulfo-NHS is pure N-hydroxysulfosuccinimide, the essential compound needed to prepare water-soluble, amine-reactive NHS-ester crosslinkers or to enhance conjugation efficiencies when using the carbodiimide EDC.
Highlights:
- Forms an acyl amino ester with carboxyl groups when used with the EDC
- Sulfo-NHS-activated molecule reacts efficiently with primary amines, resulting in stable amide bond
- Allows for two-step EDC reactions between carboxyl- and amine-containing molecules
- Sulfo-NHS-activated molecule can be stabilized by drying, allowing manufacture of amine-reactive probes and surfaces
- Reactive groups: Sulfo-NHS
- Reactive toward: amino groups (upon activation of carboxylates)

Specifications
Specifications
Chemical Reactivity | Amine-Carboxyl |
Cleavable | No |
Description | Sulfo-NHS |
Molecular Weight (g/mol) | 217.13 |
PEGylated | No |
Content And Storage | Upon receipt store at 4°C. |
Cell Permeability | No |
Shipping Condition | Ambient |
Product Line | Pierce™ |
Labeling Method | Chemical Labeling |
Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction