Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Triethylamine (TEA), Sequencing Grade PROMO

Catalog No. PI25108PM
Change view
Click to view available options
Quantity:
100 g
Catalog No. Quantity
PI25108PM 100 g
1 options

Catalog No. PI25108PM

Supplier: Thermo Scientific™ 25108

Bottles of 99.5%-pure TEA (MW 101.2, CAS 121-44-8) liquid (sold by weight); for HPLC use as an ion-pairing agent in reverse-phase peptide separations.

Triethylamine is an ion-pairing reagent that alters selectivity in reverse-phase HPLC separations. By pairing with peptides, it effectively sharpens peaks, resulting in improved peak resolution.

Highlights:

  • >99.5% triethylamine purity, allowing sensitive peptide detection at low UV wavelengths in reverse-phase HPLC peptide separation systems Packaged in amber glass bottles with protective PTFE-lined fluorocarbon caps for reagent integrity
  • Has a low UV absorbance to provide the most sensitive detection across all wavelengths

Properties of Triethylamine

  • Alternate names TEA, Diethylethanamine
  • Molecular formula C6H15N
  • Molecular weight 101.19
  • Density 0.726g/mL
  • CAS: 121-44-8

Ordering Information:

X in the ordering table indicates that hazardous shipping charges apply.

Specifications

Product Line Pierce
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.
Form Liquid
Reagent Type Ion Pairing Reagent
Quantity 100 g
Type Triethylamine (TEA)

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.