Learn More
Thermo Scientific™ Pierce™ Trifluoroacetic Acid (TFA), LC-MS Grade PROMO
Description
For complex peptide separations, the key to success can be to vary selectivity. Varying mobile phase composition on the same column can change selectivity enough to resolve peptide that would otherwise overlap. The TFA concentration is usually specified as 0.1% for reverse-phase HPLC of peptides. For reproducible separations from run-to-run or from lab-to-lab, it is essential to make concentrations the same.
Highlights:
- Purity: >99.5% TFA and exceptional clarity allows sensitive, nondestructive peptide detection at low UV wavelengths
- High-performance packaging: Packaged under nitrogen in amber glass with protective TFE-lined fluorocarbon caps to ensure TFA integrity
- Choice of formats for convenience: 1 mL ampules can prepare 1L of 0.1% v/v TFA solution for the mobile phase in reverse-phase chromatography in moments
Ordering Information:
X in the ordering table indicates that hazardous shipping charges apply.
Specifications
Specifications
| Item Name | Trifluoroacetic Acid (TFA) |
| Content And Storage | Store in a cool, dry, and well-ventilated area. Protect from direct sunlight. |
| Form | Liquid |
| Reagent Type | Ion Pairing Reagent |
| Quantity | 50 mL |
| Formulation | 99.5% TFA |
| Product Line | Pierce |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.